Assay Details
Target Gene Details
Entrez Gene ID: | 17476 |
Gene Name: | macrophage expressed gene 1 |
Gene Aliases: |
MPS1, Mpg-1 |
Location: |
Chr.19:12460709-12465285 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mpeg1 | NM_010821.1 | 2 | 2209 | NP_034951.1 |
AK030216.1 | 2 | 2210 | ||
BC049256.1 | 1 | 516 | AAH49256.1 | |
BC129843.1 | 1 | 2103 | ||
BC129844.1 | 1 | 2103 | ||
L20315.1 | 1 | 2112 | AAA73957.1 |