Assay Details
Target Gene Details
Entrez Gene ID: | 67869 |
Gene Name: | polyadenylate-binding protein-interacting protein 2 |
Gene Aliases: |
2310050K10Rik, AU045972 |
Location: |
Chr.18:35598602-35617185 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Paip2 | NM_026420.2 | NP_080696.1 | ||
AK005462.1 | BAB24053.1 | |||
AK008252.1 | ||||
AK009909.1 | BAB26578.1 | |||
AK146327.1 | BAE27082.1 | |||
AK150957.1 | BAE29989.1 | |||
AK154453.1 | BAE32597.1 | |||
AK167545.1 | BAE39611.1 | |||
AK169207.1 | BAE40979.1 | |||
AK169411.1 | BAE41157.1 | |||
BC010190.1 | AAH10190.1 |
Target Gene Details
Entrez Gene ID: | 20522 |
Gene Name: | solute carrier family 23 (nucleobase transporters), member 1 |
Gene Aliases: |
D18Ucla2, SVCT1, Slc23a2, YSPL3 |
Location: |
Chr.18:35604041-35630340 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc23a1 | NM_011397.4 | 15 | 2290 | NP_035527.3 |
AK085499.1 | 15 | 2205 | BAC39457.1 | |
AK143959.1 | 15 | 2291 | BAE25631.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |