Assay Details
Target Gene Details
Entrez Gene ID: | 107250 |
Gene Name: | Kazal-type serine peptidase inhibitor domain 1 |
Gene Aliases: |
AI842353, Bono1, IGFBP-rP10 |
Location: |
Chr.19:45074485-45079289 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kazald1 | NM_178929.4 | 2 | 283 | NP_849260.2 |
AK030678.1 | 2 | 282 | BAC27075.1 | |
AK039647.1 | 1 | 266 | ||
AK155095.1 | 3 | 528 | BAE33042.1 | |
BB618180.1 | 2 | 282 | ||
BB871162.1 | ||||
BC115642.1 | 2 | 274 |