Assay Details
Target Gene Details
Entrez Gene ID: | 108673 |
Gene Name: | coiled-coil domain containing 86 |
Gene Aliases: |
4933411H20Rik, 6720480F16Rik, D19Ertd678e |
Location: |
Chr.19:10941481-10949266 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc86 | NM_023731.3 | 1 | 503 | NP_076220.2 |
AB041663.1 | 1 | 472 | BAA95108.1 | |
AK036812.1 | 1 | 493 | ||
AK078407.1 | 1 | 504 | BAC37260.1 | |
AK078455.1 | 1 | 504 | BAC37282.1 | |
AK088727.1 | 1 | 496 | BAC40530.1 | |
AK162676.1 | 1 | 489 | ||
BC043326.1 | 1 | 499 | AAH43326.1 | |
DQ501252.1 | 1 | 470 |