Assay Details
Target Gene Details
Entrez Gene ID: | 12333 |
Gene Name: | calpain 1 |
Gene Aliases: |
Capa-1, Capa1, mu-calpin |
Location: |
Chr.19:5988545-6015825 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Capn1 | NM_001110504.1 | 2 | 181 | NP_001103974.1 |
NM_007600.3 | 2 | 233 | NP_031626.1 | |
AF021847.1 | 2 | 112 | AAB72222.1 | |
AF084459.1 | 1 | 99 | AAC33134.1 | |
AK088547.1 | 2 | 153 | BAC40416.1 | |
AK149706.1 | 2 | 172 | ||
AK150084.1 | 2 | 172 | BAE29293.1 | |
AK160940.1 | 2 | 173 | BAE36103.1 | |
AK165258.1 | 2 | 192 | ||
AK168063.1 | 2 | 172 | BAE40039.1 | |
AK171405.1 | 2 | 136 | BAE42435.1 | |
BC026138.1 | 2 | 148 | AAH26138.1 | |
BC050276.1 | 2 | 149 | AAH50276.1 | |
BY034288.1 | 2 | 182 | ||
CJ078846.1 | 2 | 234 |
Target Gene Details
Entrez Gene ID: | 100502985 |
Gene Name: | predicted gene 10814 |
Gene Aliases: |
ENSMUSG00000075230 |
Location: |
Chr.19:6012620-6018459 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm10814 | NR_045783.1 | |||
AK160921.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |