Assay Details
Target Gene Details
Entrez Gene ID: | 106512 |
Gene Name: | G-protein signalling modulator 3 (AGS3-like, C. elegans) |
Gene Aliases: |
AA960287, AGS4 |
Location: |
Chr.17:34589806-34591754 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpsm3 | NM_134116.5 | 4 | 856 | NP_598877.1 |
AK144833.1 | 4 | 839 | ||
AK146245.1 | 4 | 847 | BAE27009.1 | |
AK155617.1 | 4 | 857 | BAE33348.1 | |
AK155863.1 | 4 | 534 | BAE33467.1 | |
BC021942.1 | 4 | 446 | AAH21942.1 | |
BC080804.1 | 5 | 559 | AAH80804.1 | |
BC085186.1 | 4 | 838 | ||
BQ031296.1 | 1 | 504 |