Assay Details
Target Gene Details
Entrez Gene ID: | 70573 |
Gene Name: | TBCC domain containing 1 |
Gene Aliases: |
5730478M09Rik, AU015083, BB165076 |
Location: |
Chr.16:22813215-22857701 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tbccd1 | NM_001081368.1 | NP_001074837.1 | ||
AK017705.1 | BAB30885.1 | |||
AK035909.1 | ||||
AK039905.1 | BAC30476.1 | |||
AK040787.1 | ||||
AK043059.1 | BAC31450.1 | |||
AK133804.1 | ||||
AK200239.1 | ||||
BC082558.1 | AAH82558.1 | |||
CF749564.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |