Assay Details
Target Gene Details
Entrez Gene ID: | 240084 |
Gene Name: | coiled-coil alpha-helical rod protein 1 |
Gene Aliases: |
Hcr |
Location: |
Chr.17:35517028-35531015 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cchcr1 | NM_146248.2 | 18 | 2600 | NP_666360.2 |
AK159780.1 | BAE35364.1 | |||
BC031416.1 | AAH31416.1 | |||
BQ031569.1 | 1 | 77 | ||
CJ238541.1 |