Assay Details
Target Gene Details
Entrez Gene ID: | 20656 |
Gene Name: | superoxide dismutase 2, mitochondrial |
Gene Aliases: |
MnSOD, Sod-2 |
Location: |
Chr.17:13007839-13018119 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sod2 | NM_013671.3 | 5 | 1765 | NP_038699.2 |
AK135435.1 | 5 | 1754 | BAE22531.1 | |
AK142713.1 | 1 | 317 |
More Information
Set Membership: |
Intragenic Exonic |