Assay Details
Target Gene Details
Entrez Gene ID: | 13872 |
Gene Name: | excision repair cross-complementing rodent repair deficiency, complementation group 3 |
Gene Aliases: |
BTF2 p89, Ercc-3, XPB |
Location: |
Chr.18:32240305-32270151 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ercc3 | NM_133658.1 | 15 | 2543 | NP_598419.1 |
AK151188.1 | 15 | 2548 | ||
AK152347.1 | 15 | 2548 | ||
AK152988.1 | 15 | 2555 | ||
AK154745.1 | 15 | 2549 | BAE32801.1 | |
AK160187.1 | 9 | 1647 | BAE35681.1 | |
BC016595.1 | 15 | 2514 | AAH16595.1 | |
BC024446.1 | 6 | 889 | AAH24446.1 | |
BC026575.1 | 15 | 2525 | AAH26575.1 | |
S71186.1 | 15 | 2543 | AAB20614.1 |
More Information
Set Membership: |
Intragenic Exonic |