Assay Details
Target Gene Details
Entrez Gene ID: | 81877 |
Gene Name: | tenascin XB |
Gene Aliases: |
Tn-mhc, Tnx |
Location: |
Chr.17:34659788-34795460 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tnxb | NM_031176.2 | 5 | 2610 | NP_112453.2 |
AB235155.1 | 5 | 2610 | BAE44473.1 |
More Information
Set Membership: |
Intragenic Exonic |