Assay Details
Target Gene Details
Entrez Gene ID: | 65962 |
Gene Name: | solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 2 |
Gene Aliases: |
0610011L07Rik, 1200011K07Rik, 2010007A20Rik, E3karp, NHERF-2, Nherf2, Octs2, Sip-1, Sip1, Sryip1, Tka-1 |
Location: |
Chr.17:24639281-24650340 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc9a3r2 | NM_023055.2 | 7 | 1939 | NP_075542.2 |
NM_023449.3 | 6 | 1509 | NP_075938.2 | |
AB026489.1 | 7 | 1912 | BAA97568.1 | |
AK004710.1 | 3 | 1182 | ||
AK039491.1 | 7 | 1927 | BAC30366.1 | |
AK084801.1 | 7 | 1940 | BAC39282.1 | |
AK170057.1 | 7 | 1925 | BAE41537.1 | |
BC065778.1 | AAH65778.1 | |||
BC070458.1 | 6 | 1860 | AAH70458.1 | |
CJ194744.1 | 1 | 188 |