Assay Details
Target Gene Details
Entrez Gene ID: | 73473 |
Gene Name: | IWS1 homolog (S. cerevisiae) |
Gene Aliases: |
1700069O15Rik, 3000008H23, AL117927, AW214353 |
Location: |
Chr.18:32067325-32104331 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Iws1 | NM_173441.3 | 1 | 44 | NP_775617.1 |
AK028247.1 | BAC25838.1 | |||
AK033013.1 | 1 | 39 | ||
AK041691.1 | 1 | 38 | BAC31034.1 | |
AK163972.1 | BAE37560.1 | |||
BC068184.1 | AAH68184.1 |