Assay Details
Target Gene Details
Entrez Gene ID: | 213012 |
Gene Name: | abhydrolase domain containing 10 |
Gene Aliases: |
- |
Location: |
Chr.16:45729720-45743278 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abhd10 | NM_001272070.1 | NP_001258999.1 | ||
NM_172511.4 | 5 | 881 | NP_766099.3 | |
NR_073572.1 | 5 | 739 | ||
AK039713.1 | 1 | 2110 | ||
AK051923.1 | 1 | 4070 | ||
AK052670.1 | 5 | 830 | BAC35091.1 | |
AK084708.1 | 1 | 3451 | ||
AK084998.1 | BAC39335.1 | |||
AK166025.1 | 2 | 307 | BAE38528.1 | |
AK187201.1 | ||||
AK211815.1 | ||||
BC027656.1 | 2 | 296 | AAH27656.1 | |
BC058347.1 | 5 | 830 | AAH58347.1 | |
CX237340.1 | 5 | 686 |
More Information
Set Membership: |
Intragenic Exonic |