Assay Details
Target Gene Details
Entrez Gene ID: | 74374 |
Gene Name: | C-type lectin domain family 16, member A |
Gene Aliases: |
4932416N17Rik, curt, mKIAA0350 |
Location: |
Chr.16:10545339-10744878 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clec16a | NM_001204229.1 | 23 | 5695 | NP_001191158.1 |
NM_177562.5 | 24 | 5701 | NP_808230.2 | |
AK028051.1 | 1 | 1955 | ||
AK046244.1 | 1 | 807 | ||
AK122255.1 | 24 | 5653 | BAC65537.1 |