Assay Details
Target Gene Details
Entrez Gene ID: | 106014 |
Gene Name: | family with sequence similarity 19, member A5 |
Gene Aliases: |
TAFA5, Tafa-5, Tara-5 |
Location: |
Chr.15:87544299-87759364 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam19a5 | NM_001252310.1 | 4 | 1957 | NP_001239239.1 |
NM_134096.2 | 4 | 1909 | NP_598857.1 | |
NR_045510.1 | 3 | 1635 | ||
AK090307.1 | 4 | 1959 | ||
BC015306.1 | 4 | 1910 | AAH15306.1 |
More Information
Set Membership: |
Intragenic Exonic |