Assay Details
Target Gene Details
Entrez Gene ID: | 72555 |
Gene Name: | shisa family member 9 |
Gene Aliases: |
2700045P11Rik, Ckamp44 |
Location: |
Chr.16:11984113-12270904 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Shisa9 | NM_001174086.1 | 1 | 858 | NP_001167557.1 |
NM_028277.2 | 1 | 858 | NP_082553.2 | |
AK012380.1 | 1 | 859 | BAB28201.1 | |
AK049860.1 | 1 | 384 | ||
GU479981.1 | 1 | 390 | ||
GU479982.1 | 1 | 390 |