Assay Details
Target Gene Details
Entrez Gene ID: | 105446 |
Gene Name: | guanosine monophosphate reductase 2 |
Gene Aliases: |
1810008P16Rik, 5730544D12Rik, AA959850 |
Location: |
Chr.14:55672235-55687634 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gmpr2 | NM_177992.2 | 10 | 1364 | NP_818773.1 |
AK031004.1 | 10 | 1366 | BAC27211.1 | |
AK075772.1 | 10 | 1322 | BAC35946.1 | |
BC003886.1 | 10 | 1323 | AAH03886.1 | |
BC024109.1 | 10 | 1320 | AAH24109.1 | |
BE956837.1 | 1 | 197 |
Target Gene Details
Entrez Gene ID: | 28113 |
Gene Name: | Terf1 (TRF1)-interacting nuclear factor 2 |
Gene Aliases: |
AW552114, D14Wsu146e, Tin2 |
Location: |
Chr.14:55676208-55682624 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tinf2 | AK028952.1 | 8 | 2373 | |
AK144592.1 | BAE25954.1 | |||
AK169480.1 | 3 | 2365 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |