Assay Details
Target Gene Details
Entrez Gene ID: | 20512 |
Gene Name: | solute carrier family 1 (glial high affinity glutamate transporter), member 3 |
Gene Aliases: |
AI504299, B430115D02Rik, Eaat1, GLAST, GLAST-1, GLU-T, GluT-1, Gmt1, MGluT1 |
Location: |
Chr.15:8634124-8710807 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc1a3 | NM_148938.3 | 10 | 3837 | NP_683740.1 |
AK031831.1 | 10 | 3500 | ||
AK080879.1 | 10 | 3521 | ||
AK082736.1 | 10 | 3497 | BAC38592.1 | |
BC058711.1 | 10 | 3760 | AAH58711.1 |
More Information
Set Membership: |
Intragenic Exonic |