Assay Details
Target Gene Details
Entrez Gene ID: | 20491 |
Gene Name: | src-like adaptor |
Gene Aliases: |
Slap, Slap-1 |
Location: |
Chr.15:66780819-66831829 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sla | NM_001029841.4 | 8 | 1876 | NP_001025012.1 |
NM_009192.3 | 9 | 2145 | NP_033218.1 | |
AJ131777.1 | 8 | 1783 | CAB66139.1 | |
AK036167.1 | 8 | 1877 | BAC29328.1 | |
AK037901.1 | 8 | 1787 | BAC29896.1 | |
AK137954.1 | 8 | 1811 | BAE23522.1 | |
BC032922.1 | 8 | 1681 | AAH32922.1 |
Target Gene Details
Entrez Gene ID: | 21819 |
Gene Name: | thyroglobulin |
Gene Aliases: |
Tgn, cog |
Location: |
Chr.15:66670756-66850721 on Build GRCm38 |
Assay Gene Location: | Within Intron 41 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tg | NM_009375.2 | NP_033401.2 | ||
AF076186.1 | AAC32268.1 | |||
AF076187.1 | AAC32269.1 | |||
BC111467.1 | AAI11468.1 | |||
U76389.1 | AAB53204.1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |