Assay Details
Target Gene Details
Entrez Gene ID: | 238393 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A, member 3F |
Gene Aliases: |
2A1, BC049975 |
Location: |
Chr.12:104214544-104221129 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Serpina3f | NM_001033335.3 | 6 | 1584 | NP_001028507.2 |
NM_001168294.1 | 6 | 1625 | NP_001161766.1 | |
NM_001168295.1 | 5 | 1413 | NP_001161767.1 | |
AK138954.1 | 6 | 1585 | BAE23834.1 | |
BC049975.1 | 5 | 1403 | AAH49975.1 | |
BC139132.1 | 5 | 1473 | ||
BC139133.1 | 5 | 1473 |
More Information
Set Membership: |
Intragenic Exonic |