Assay Details
Target Gene Details
Entrez Gene ID: | 271047 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade A, member 3B |
Gene Aliases: |
6A1, A030003A19Rik |
Location: |
Chr.12:104127531-104139546 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Serpina3b | NM_173024.3 | 5 | 1598 | NP_766612.1 |
AK037160.1 | 5 | 1600 | BAC29726.1 | |
BC125418.1 | 5 | 1581 | ||
BC145808.1 | 5 | 1581 |
More Information
Set Membership: |
Intragenic Exonic |