Assay Details
Target Gene Details
Entrez Gene ID: | 14186 |
Gene Name: | fibroblast growth factor receptor 4 |
Gene Aliases: |
Fgfr-4 |
Location: |
Chr.13:55152818-55168759 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fgfr4 | NM_008011.2 | 18 | 2658 | NP_032037.2 |
AK084850.1 | 18 | 2658 | BAC39292.1 | |
AK085723.1 | 18 | 2656 | ||
AY493377.2 | 16 | 2406 | AAS72387.2 | |
BC033313.1 | 18 | 2845 | AAH33313.1 | |
DQ388428.1 | 17 | 2544 | ABD43187.1 | |
X59927.1 | 18 | 2627 | CAA42551.1 |
More Information
Set Membership: |
Intragenic Exonic |