Assay Details
Target Gene Details
Entrez Gene ID: | 57266 |
Gene Name: | chemokine (C-X-C motif) ligand 14 |
Gene Aliases: |
1110031L23Rik, 1200006I23Rik, AI414372, BMAC, BRAK, KS1, Kec, MIP-2g, MIP2gamma, NJAC, Scyb14, bolekine |
Location: |
Chr.13:56288643-56296792 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cxcl14 | NM_019568.2 | 1 | 154 | NP_062514.2 |
AB041614.1 | 1 | 154 | BAA95097.1 | |
AF144754.1 | AAF66694.1 | |||
AF192557.1 | 1 | 154 | AAF03753.1 | |
AF252873.1 | 1 | 154 | AAF65570.1 | |
AK004615.1 | BAB23411.1 | |||
AK076112.1 | BAC36192.1 | |||
AK155356.1 | BAE33213.1 | |||
AU035952.1 | 1 | 154 | ||
BI133967.1 | 1 | 245 |
More Information
Set Membership: |
Intragenic Exonic |