Assay Details
Target Gene Details
Entrez Gene ID: | 12919 |
Gene Name: | corticotropin releasing hormone binding protein |
Gene Aliases: |
CRH-BP |
Location: |
Chr.13:95431376-95444831 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Crhbp | NM_198408.3 | 7 | 1332 | NP_940800.1 |
AI854101.1 | 1 | 366 | ||
AK135067.1 | 7 | 1333 | BAE22406.1 | |
AK139404.1 | 7 | 1178 | ||
AK164173.1 | 1 | 342 | BAE37662.1 | |
BC061247.1 | 7 | 1273 | AAH61247.1 |