Assay Details
Target Gene Details
Entrez Gene ID: | 338320 |
Gene Name: | melanoma inhibitory activity 2 |
Gene Aliases: |
- |
Location: |
Chr.12:59095800-59109130 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mia2 | NM_177321.2 | 4 | 979 | NP_796295.1 |
AF390177.2 | 4 | 979 | AAL26992.2 |
More Information
Set Membership: |
Intragenic Exonic |