Assay Details
Target Gene Details
Entrez Gene ID: | 68675 |
Gene Name: | family with sequence similarity 172, member A |
Gene Aliases: |
1110033M05Rik, 2610318O14Rik, 53-E6, 9430037D06Rik, AF064782, pEN87 |
Location: |
Chr.13:77708678-78166240 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam172a | NM_001163419.1 | 10 | 3431 | NP_001156891.1 |
NM_001163420.1 | 9 | 3239 | NP_001156892.1 | |
NM_138312.1 | 11 | 3555 | NP_612185.1 | |
NR_028109.1 | 11 | 3480 | ||
AK052419.1 | 10 | 3404 | ||
AW060304.1 | 1 | 482 | ||
BC057380.1 | 10 | 3399 | AAH57380.1 |