Assay Details
Target Gene Details
Entrez Gene ID: | 26363 |
Gene Name: | biotinidase |
Gene Aliases: |
- |
Location: |
Chr.14:31640999-31668197 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Btd | NM_025295.4 | 4 | 1446 | NP_079571.1 |
AK005506.1 | 4 | 1439 | BAB24086.1 | |
AK135375.1 | 4 | 1436 | BAE22509.1 | |
BC024051.1 | 4 | 1413 | ||
BU700769.1 | 1 | 511 |
More Information
Set Membership: |
Intragenic Exonic |