Assay Details
Target Gene Details
Entrez Gene ID: | 217692 |
Gene Name: | signal-induced proliferation-associated 1 like 1 |
Gene Aliases: |
4931426N11Rik, AW213287, Spar, mKIAA0440 |
Location: |
Chr.12:82169378-82451786 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sipa1l1 | NM_001167983.1 | 12 | 3614 | NP_001161455.1 |
NM_172579.3 | 9 | 3418 | NP_766167.2 | |
AK029889.1 | 10 | 3444 | BAC26660.1 | |
AK051853.1 | 8 | 1752 | ||
AK122284.1 | 8 | 2202 | BAC65566.1 | |
AK147591.1 | 11 | 4139 | ||
AK171876.1 | BAE42714.1 | |||
BC058681.1 | 12 | 3604 | AAH58681.1 | |
BC095958.1 | 12 | 3603 | AAH95958.1 |
More Information
Set Membership: |
Intragenic Exonic |