Assay Details
Target Gene Details
Entrez Gene ID: | 11298 |
Gene Name: | arylalkylamine N-acetyltransferase |
Gene Aliases: |
AA-NAT, Nat-2, Nat4, Snat |
Location: |
Chr.11:116591721-116597680 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aanat | NM_009591.3 | 4 | 836 | NP_033721.1 |
NR_033223.1 | 5 | 938 | ||
AF004108.1 | 4 | 836 | AAD09408.1 | |
AF004109.1 | 3 | 825 | AAD09409.1 | |
AF004110.1 | 5 | 938 | AAD09410.1 | |
AF004111.1 | 4 | 850 | AAD09411.1 | |
BC116967.1 | 4 | 807 | ||
BC119139.1 | 4 | 807 | ||
CO040083.1 | 1 | 535 |
More Information
Set Membership: |
Intragenic Exonic |