Assay Details
Target Gene Details
Entrez Gene ID: | 216877 |
Gene Name: | DEAH (Asp-Glu-Ala-His) box polypeptide 33 |
Gene Aliases: |
3110057P17Rik, 9430096J02Rik, Ddx33 |
Location: |
Chr.11:70984091-71004553 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dhx33 | NM_178367.4 | 12 | 4624 | NP_848144.3 |
AK014230.1 | 1 | 1371 | ||
BC052172.1 | 12 | 4618 | AAH52172.1 | |
BQ287292.1 | 1 | 457 |
More Information
Set Membership: |
Intragenic Exonic |