Assay Details
Target Gene Details
Entrez Gene ID: | 103677 |
Gene Name: | Smg-6 homolog, nonsense mediated mRNA decay factor (C. elegans) |
Gene Aliases: |
AI317223, AU041178 |
Location: |
Chr.11:74925872-75164448 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smg6 | NM_001002764.1 | 19 | 5528 | NP_001002764.1 |
AK032934.1 | 1 | 331 | ||
AK052642.1 | 2 | 1634 | ||
AK129203.1 | 18 | 4147 | BAC98013.1 | |
AK145229.1 | 3 | 2014 | ||
BC066040.1 | 19 | 5528 | AAH66040.1 |
More Information
Set Membership: |
Intragenic Exonic |