Assay Details
Target Gene Details
Entrez Gene ID: | 13855 |
Gene Name: | epsin 2 |
Gene Aliases: |
9530051D10Rik, AA536924, Ibp2 |
Location: |
Chr.11:61517249-61579881 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Epn2 | NM_001252188.1 | 11 | 2408 | NP_001239117.1 |
NM_001252189.1 | 9 | 2164 | NP_001239118.1 | |
NM_010148.3 | 10 | 2200 | NP_034278.1 | |
AF057286.1 | 8 | 1655 | AAC97476.1 | |
AK036331.1 | 9 | 2138 | BAC29387.1 | |
AK139234.1 | 11 | 2396 | ||
AK139793.1 | 11 | 2331 | ||
AK160782.1 | 9 | 2165 | BAE36006.1 | |
AK173093.1 | 11 | 2384 | BAD32371.1 | |
BC039138.1 | 10 | 2164 | AAH39138.1 |