Assay Details
Target Gene Details
Entrez Gene ID: | 52626 |
Gene Name: | CDKN2A interacting protein N-terminal like |
Gene Aliases: |
A430101B06Rik, AA409260, D11Ertd497e, MNCb-1520 |
Location: |
Chr.11:51967631-51977336 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdkn2aipnl | NM_029976.3 | 1 | 135 | NP_084252.1 |
AB041653.1 | 1 | 103 | BAB93546.1 | |
AK020751.1 | 1 | 108 | BAB32201.1 | |
AK077515.1 | ||||
AK150573.1 | 1 | 91 | BAE29668.1 | |
AK171740.1 | 1 | 70 | BAE42643.1 | |
BC116362.2 | 1 | 95 | ||
BC116363.2 | 1 | 95 | ||
BY088687.1 | 1 | 136 |
More Information
Set Membership: |
Intragenic Exonic |