Assay Details
Target Gene Details
Entrez Gene ID: | 14708 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma 7 |
Gene Aliases: |
AI840417 |
Location: |
Chr.10:80948624-81014961 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gng7 | NM_001038655.1 | 4 | 2384 | NP_001033744.1 |
NM_010319.3 | 4 | 2321 | NP_034449.2 | |
AK141541.1 | 4 | 2293 | BAE24725.1 | |
AK147843.1 | 4 | 2284 | BAE28176.1 | |
AK148026.1 | 1 | 2056 | BAE28296.1 | |
AK158963.1 | 4 | 2384 | BAE34744.1 | |
BC034108.1 | 3 | 2248 | AAH34108.1 |
More Information
Set Membership: |
Intragenic Exonic |