Assay Details
Target Gene Details
Entrez Gene ID: | 20509 |
Gene Name: | solute carrier family 19 (folate transporter), member 1 |
Gene Aliases: |
AI323572, RFC, RFC-1, RFC1 |
Location: |
Chr.10:77032269-77050432 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc19a1 | NM_001199271.1 | 6 | 1657 | NP_001186200.1 |
NM_031196.3 | 6 | 1751 | NP_112473.1 | |
AK002560.1 | 6 | 1668 | BAB22188.1 | |
AK079592.1 | 6 | 1712 | BAC37693.1 | |
AK085024.2 | 6 | 1699 | ||
AK088803.1 | 6 | 1714 | BAC40582.1 | |
AK138577.1 | 6 | 1577 | ||
BC015263.1 | 6 | 1669 | AAH15263.1 | |
L23755.1 | 5 | 1609 | AAA39738.1 | |
L36539.1 | 6 | 1664 | AAB38483.1 | |
U32469.1 | 6 | 1691 | AAC52258.1 | |
U66103.1 | 6 | 1704 | AAC53287.1 |
More Information
Set Membership: |
Intragenic Exonic |