Assay Details
Target Gene Details
Entrez Gene ID: | 15586 |
Gene Name: | hyaluronoglucosaminidase 1 |
Gene Aliases: |
Hya1, Hyal-1 |
Location: |
Chr.9:107576952-107580137 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hyal1 | NM_008317.4 | 4 | 1308 | NP_032343.2 |
AF011567.1 | 4 | 1294 | AAC15949.1 | |
AF422176.1 | 4 | 1256 | AAL17822.1 | |
BC021636.1 | 4 | 1309 | AAH21636.1 |
More Information
Set Membership: |
Intragenic Exonic |