Assay Details
Target Gene Details
Entrez Gene ID: | 74032 |
Gene Name: | short chain dehydrogenase/reductase family 42E, member 1 |
Gene Aliases: |
4632417N05Rik, AI117603 |
Location: |
Chr.8:117661399-117673718 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sdr42e1 | NM_028725.3 | 1 | 254 | NP_083001.1 |
AK014586.1 | 1 | 256 | BAB29446.1 | |
BC038819.1 | AAH38819.1 |
More Information
Set Membership: |
Intragenic Exonic |