Assay Details
Target Gene Details
Entrez Gene ID: | 244349 |
Gene Name: | K(lysine) acetyltransferase 6A |
Gene Aliases: |
1500036M03, 9930021N24Rik, MOZ, Myst3, Zfp220 |
Location: |
Chr.8:22859516-22943262 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kat6a | NM_001081149.1 | 16 | 3331 | NP_001074618.1 |
AK036885.1 | 16 | 3326 | BAC29621.1 | |
AK082463.1 | 16 | 3334 | ||
AK132474.1 | 15 | 3239 | BAE21185.1 | |
AK143408.1 | 16 | 3322 | BAE25366.1 | |
BC024786.1 | 8 | 1457 |
More Information
Set Membership: |
Intragenic Exonic |