Assay Details
Target Gene Details
Entrez Gene ID: | 72254 |
Gene Name: | RIKEN cDNA 1700030K09 gene |
Gene Aliases: |
- |
Location: |
Chr.8:72443880-72460541 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1700030K09Rik | NM_028170.2 | 5 | 1499 | NP_082446.2 |
AK089918.1 | 5 | 1441 | ||
AK133815.1 | 5 | 1503 | BAE21858.1 | |
AK157359.1 | 4 | 710 | ||
AK172202.1 | 4 | 708 | ||
BC008558.1 | 5 | 1453 | AAH08558.1 |