Assay Details
Target Gene Details
Entrez Gene ID: | 244853 |
Gene Name: | neurexophilin and PC-esterase domain family, member 4 |
Gene Aliases: |
C130036J11, D930028F11Rik, Fam55d |
Location: |
Chr.9:48162001-48400025 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nxpe4 | NM_172921.3 | 6 | 801 | NP_766509.2 |
AK048132.1 | 7 | 813 | BAC33253.1 | |
AK136478.1 | 2 | 195 | BAE22996.1 | |
BC085195.1 | 2 | 624 | ||
BC094249.1 | 8 | 1000 | AAH94249.1 |