Assay Details
Target Gene Details
Entrez Gene ID: | 12768 |
Gene Name: | chemokine (C-C motif) receptor 1 |
Gene Aliases: |
Cmkbr1, Mip-1a-R |
Location: |
Chr.9:123962126-123968692 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccr1 | NM_009912.4 | 1 | 199 | NP_034042.3 |
AK031109.1 | 1 | 200 | BAC27256.1 | |
AK036690.1 | 1 | 200 | BAC29535.1 | |
AK076275.1 | 1 | 74 | BAC36284.1 |