Assay Details
Target Gene Details
Entrez Gene ID: | 232943 |
Gene Name: | kinesin light chain 3 |
Gene Aliases: |
BC017147, Klct |
Location: |
Chr.7:19394440-19404442 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klc3 | NM_001286038.1 | 2 | 386 | NP_001272967.1 |
NM_001286039.1 | 2 | 262 | NP_001272968.1 | |
NM_146182.4 | 2 | 291 | NP_666294.1 | |
AK158094.1 | 2 | 228 | BAE34354.1 | |
BC017147.1 | 2 | 228 | AAH17147.1 | |
BY016862.1 | 2 | 291 | ||
BY127751.1 | 2 | 263 |
More Information
Set Membership: |
Intragenic Exonic |