Assay Details
Target Gene Details
Entrez Gene ID: | 233899 |
Gene Name: | predicted gene 166 |
Gene Aliases: |
- |
Location: |
Chr.7:127582383-127588609 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm166 | NM_001033040.3 | 9 | 1750 | NP_001028212.1 |
AK140121.1 | 9 | 1751 | BAE24243.1 | |
BC066021.1 | 9 | 1331 |
Target Gene Details
Entrez Gene ID: | 68961 |
Gene Name: | phosphorylase kinase, gamma 2 (testis) |
Gene Aliases: |
1500017I02Rik |
Location: |
Chr.7:127573344-127583307 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Phkg2 | NM_026888.3 | 10 | 1488 | NP_081164.2 |
AK005277.1 | 10 | 1433 | BAB23926.1 | |
BC138913.1 | 10 | 1402 | ||
BC145734.1 | 10 | 1402 | ||
BQ176174.1 | 1 | 157 |
More Information
Set Membership: |
Intragenic Exonic |