Assay Details
Target Gene Details
Entrez Gene ID: | 654804 |
Gene Name: | RIKEN cDNA 4732471J01 gene |
Gene Aliases: |
- |
Location: |
Chr.7:25376819-25566417 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4732471J01Rik | NR_015569.3 | 1 | 939 | |
NR_073027.1 | 2 | 991 | ||
AK043644.1 | 2 | 989 | ||
AK076372.1 | 2 | 991 | ||
AK161422.1 | 1 | 239 |
Target Gene Details
Entrez Gene ID: | 16890 |
Gene Name: | lipase, hormone sensitive |
Gene Aliases: |
4933403G17Rik, HSL |
Location: |
Chr.7:25379527-25398704 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lipe | NM_010719.5 | 1 | 337 | NP_034849.2 |
AK019846.1 | 1 | 730 | BAB31878.1 | |
AK029984.1 | 1 | 340 | BAC26716.1 |
More Information
Set Membership: |
Intragenic Exonic |