Assay Details
Target Gene Details
Entrez Gene ID: | 78829 |
Gene Name: | TSC22 domain family, member 4 |
Gene Aliases: |
0610009M14Rik, 1700023B23Rik, AI415410, Thg-1pit, Tilz2 |
Location: |
Chr.5:137745969-137768453 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tsc22d4 | NM_023910.6 | 1 | 240 | NP_076399.4 |
NM_029805.1 | 1 | 240 | NP_084081.1 | |
AK018735.2 | 1 | 213 | BAB31377.1 | |
AK039535.1 | 1 | 198 | ||
AK040534.1 | 1 | 177 | BAC30619.1 | |
AK080828.1 | 1 | 251 | ||
AK143608.1 | BAE25460.1 | |||
AK150043.1 | 1 | 211 | ||
AK154153.1 | 1 | 190 | BAE32411.1 | |
AK155712.1 | 1 | 240 | BAE33397.1 | |
AK170487.1 | 1 | 229 | BAE41829.1 | |
BC023761.1 | 1 | 144 | AAH23761.1 | |
BC145721.1 | 1 | 203 | ||
BC145723.1 | 1 | 203 | ||
BY750078.1 | 1 | 240 |