Assay Details
Target Gene Details
Entrez Gene ID: | 109305 |
Gene Name: | ORAI calcium release-activated calcium modulator 1 |
Gene Aliases: |
D730049H07Rik, Tmem142a, orai-1 |
Location: |
Chr.5:123015074-123030452 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Orai1 | NM_175423.3 | 2 | 1417 | NP_780632.1 |
AK052573.1 | BAC35045.1 | |||
AK085755.1 | 2 | 1168 | BAC39533.1 | |
BC023149.1 | 2 | 1189 | AAH23149.1 | |
BM223639.2 |
More Information
Set Membership: |
Intragenic Exonic |