Assay Details
Target Gene Details
Entrez Gene ID: | 20392 |
Gene Name: | sarcoglycan, epsilon |
Gene Aliases: |
e-SG |
Location: |
Chr.6:4674350-4747204 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sgce | NM_001130188.1 | 12 | 1662 | NP_001123660.1 |
NM_001130189.1 | 11 | 1562 | NP_001123661.1 | |
NM_001130190.1 | 12 | 1625 | NP_001123662.1 | |
NM_001130191.1 | 11 | 1598 | NP_001123663.1 | |
NM_011360.3 | 11 | 1554 | NP_035490.3 | |
AB117975.1 | BAD21207.1 | |||
AB200281.1 | 12 | 1440 | BAE16175.1 | |
AB200282.1 | 11 | 1413 | BAE16176.1 | |
AF031919.1 | 11 | 1410 | AAC14020.1 | |
AF103877.1 | 12 | 1468 | AAF21895.1 | |
AK076102.1 | 11 | 1376 | BAC36184.1 | |
AK215752.1 | 1 | 153 | ||
AW457608.1 | 1 | 158 | ||
BC012665.1 | 11 | 1460 | AAH12665.1 |
More Information
Set Membership: |
Intragenic Exonic |