Assay Details
Target Gene Details
Entrez Gene ID: | 232164 |
Gene Name: | poly(A) binding protein interacting protein 2B |
Gene Aliases: |
- |
Location: |
Chr.6:83806058-83831741 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Paip2b | NM_146169.2 | 5 | 1447 | NP_666281.2 |
AK159287.1 | 5 | 1448 | BAE34963.1 | |
AK168804.1 | 5 | 1396 | BAE40634.1 | |
AK173114.1 | 5 | 1389 | BAD32392.1 | |
BC017133.1 | 5 | 1370 | AAH17133.2 | |
BC034096.1 | 5 | 1395 | AAH34096.2 | |
BC039969.1 | 5 | 1406 | ||
BC147239.1 | 4 | 1342 | ||
BC147240.1 | 4 | 1347 |