Assay Details
Target Gene Details
Entrez Gene ID: | 66506 |
Gene Name: | proteasome (prosome, macropain) assembly chaperone 3 |
Gene Aliases: |
1810042K04Rik, 4930403H09Rik, AI303239 |
Location: |
Chr.5:139823594-139827003 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Psmg3 | NM_025604.3 | 3 | 764 | NP_079880.1 |
AK012616.1 | 3 | 765 | BAB28357.1 | |
BC027410.1 | AAH27410.1 | |||
BC058692.1 | AAH58692.1 |
More Information
Set Membership: |
Intragenic Exonic |